Learn More
Sino Biological Anti-TARC/CCL17 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10233MM08H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCL17/TARC (rh CCL17/TARC; Catalog#10233-HNAB; Q92583-1; Met1-Ser94). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8503