Learn More
Sino Biological Recombinant Anti-CCL18/PARC/MIP-4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10502R016100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL18 / PARC / MIP-4 (rh CCL18 / PARC / MIP-4; Catalog#10502-H08Y; NP_002979.1; Ala21-Ala89).
Catalog No. 50-240-9212