Learn More
Sino Biological Recombinant Anti-MCP-1/CCL2 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 10134R144H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human MCP-1 / CCL2 (rh MCP-1 / CCL2; Catalog#10134-H08Y; NP_002973.1; Gln24-Thr99), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-1623