Learn More
Sino Biological Recombinant Anti-MCP-1/CCL2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10134R12320
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human MCP-1/CCL2 (rh MCP-1/CCL2; Catalog#10134-H08Y; NP_002973.1; Gln24-Thr99).
Catalog No. 50-241-0638