Learn More
Sino Biological Human CCL20 MAb 100UL 10485-MM21-100
Supplier:  Sino Biological 10485MM21100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCL20/MIP-3 alpha/MIP3A (rh CCL20/MIP-3 alpha/MIP3A; Catalog#10485-H07E; P78556-1; Ala27-Met96). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2191