Learn More
Sino Biological Recombinant Human CCL20/MIP-3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10485H07E1
Encompass_Preferred
A DNA sequence encoding the human CCL20 (P78556-1) (Ala27-Met96) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-5276