Learn More
Bon Opus Biosciences Recombinant Human CCL22 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP108650UG
Encompass_Preferred
Recombinant Human CCL22 (C-hFc) is produced by Human Cells. The target gene encoding Gly25-Gln93 is expressed with a C-hFc tag. Accession# AAH27952.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1086
Catalog No. 50-217-2684