Learn More
Sino Biological Recombinant Anti-MPIF-1/CCL23 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10655R007100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL23 / MIP 3 (rh CCL23 / MIP 3; Catalog#10655-H07E; AAB51134.1; Arg22-Asn120).
Catalog No. 50-241-2312