Learn More
Sino Biological Anti-CCL27 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10625MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCL27 (rh CCL27; Catalog#10625-H07E; Q9Y4X3; Phe25-Gly112). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8914