Learn More
ACROBiosystems Human CCL27 Protein His Tag
Supplier:  ACROBiosystems CC7H5143100UG
Encompass_Preferred
Human CCL27 Protein His Tag (CC7-H5143) is expressed from E. coli cells. It contains AA Phe 25 - Gly 112 (Accession # Q9Y4X3).
Catalog No. 50-253-5401