Learn More
Sino Biological Anti-CCL28 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10618MM1520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCL28 / SCYA28 (rh CCL28 / SCYA28; Catalog#10618-H07E; NP_683513.1; Ile23-Tyr127). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8608