Learn More
Sino Biological Recombinant Human MIP-1 alpha/CCL3 Protein (His Tag)
Supplier:  Sino Biological 11292H08Y1
Encompass_Preferred
A DNA sequence encoding human CCL3 (NP_002974.1) (Ala23-Ala92) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4715