Learn More
Sino Biological Human TNF-alpha / TNFA Protein (Fc Tag) 1mg
Supplier:  Sino Biological 10602H01H1
Encompass_Preferred
A DNA sequence encoding the human TNF (NP_000585.2) (Val77-Leu233) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-222-2710