Learn More
Sino Biological Anti-CCL5/RANTES Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10900MM05H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCL5 (rh CCL5; Catalog#10900-HNAE; NP_002976.2; Ser24-Ser91). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-5085