Learn More
Sino Biological Anti-CCL5/RANTES Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10900MM0520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCL5 (rh CCL5; Catalog#10900-HNAE; NP_002976.2; Ser24-Ser91). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5086