Learn More
ACROBiosystems ACROBiosystems Human CCL5/RANTES Protein, Fc Tag (MALS verified)
Catalog No. 502106515
Encompass_Preferred
Recombinant Protein;1MG;Human CCL5, Fc Tag (CC5-H5251) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 91 (Accession # P13501-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 34.4 kDa. The protein migrates as 35-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6515 Supplier ACROBiosystems Supplier No. CC5H52511MG
Recombinant Protein;1MG;Human CCL5, Fc Tag (CC5-H5251) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 91 (Accession # P13501-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 34.4 kDa. The protein migrates as 35-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.