Learn More
Sino Biological Human CCL7 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11926R063100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human MCP-3/CCL7 (rh MCP-3/CCL7; Catalog#11926-H08E; NP_006264.2; Gln24-Leu99).
Catalog No. 50-237-3303