Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CCL7 (MCP-3) Recombinant Protein, PeproTech®

Catalog No. p-7246017
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
This item is not returnable. View return policy

Catalog No. 30017100UG

Supplier: Gibco™ 30017100UG

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human MCP-3 is a 9.0 kDa protein containing 76 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This is a secreted chemokine which attracts macrophages during inflammation and metastasis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues. The protein is an in vivo substrate of matrix metalloproteinase 2, an enzyme which degrades components of the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P80098
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6354
Molecular Weight (g/mol) 9 kDa
Name Human CCL7 (MCP-3)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C-C motif chemokine 7; C-C motif chemokine ligand 7; CCL7; chemokine (C-C motif) ligand 7; chemotactic protein-3; Fic; H-MCP-3; Intercrine/chemokine MARC; marc; Mcp3; MCP-3; MGC138463; MGC138465; Monocyte chemoattractant protein 3; monocyte chemotactic protein 3; NC28; Protein FIC; RANTES/sis homolog; RP23-350G1.4; SCYA6; Scya7; small inducible cytokine A7; small inducible cytokine A7 (monocyte chemotactic protein 3); small-inducible cytokine A7
Common Name CCL7 (MCP3)
Gene Symbol CCL7
Biological Activity Determined by its ability to chemoattract human peripheral blood monocytes using a concentration range of 10.0-100.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence QPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKL
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.