Learn More
Sino Biological Recombinant Human MCP-2/CCL8 Protein (His Tag)
Supplier:  Sino Biological 10989H08Y1
Encompass_Preferred
A DNA sequence encoding the human CCL8 (NP_005614.2) (Gln24-Pro99) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4992