Learn More
Sino Biological Recombinant Human WISP1/CCN4 Protein (His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10442H08H1
Encompass_Preferred
A DNA sequence encoding the human WISP1 (NP_003873.1) (Met 1-Asn 367) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6782