Learn More
Sino Biological Human CCNA1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11013MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCNA1 (rh CCNA1; Catalog#11013-H07B; NP_003905.1; Met 1-Gln 465). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3829