Learn More
Sino Biological Anti-Cyclin D3/CCND3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101199T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Cyclin D3/CCND3, and purified by antigen affinity chromatography.
Catalog No. 50-240-8097