Learn More
Sino Biological Anti-Cyclin E Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10902MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCNE1 / Cyclin-E1 (rh CCNE1 / Cyclin-E1; Catalog#10902-H07B; NP_001229.1; Met1-Ala410). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8704