Learn More
Sino Biological Recombinant Human Cyclin E Protein (His & GST Tag)
Supplier:  Sino Biological 10902H20B100
Encompass_Preferred
A DNA sequence encoding the full length of human CCNE1 isoform 1 (NP_001229.1) (Met 1-Ala 410) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-237-1687