Learn More
Sino Biological Anti-cyclin E2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107019T02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human cyclin E2/CCNE2, and purified by antigen affinity chromatography.
Catalog No. 50-241-1606