Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CCP110 Synthetic Peptide

Catalog No. PEP1423 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1423 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1423 Supplier Invitrogen™ Supplier No. PEP1423
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 17 amino acid peptide near the carboxy terminus of human CCP110. PEP-1423 can be used as a blocking peptide with polyclonal antibody PA5-34380.

Necessary for centrosome duplication at different stages of procentriole formation. Acts as a key negative regulator of ciliogenesis in collaboration with CEP97 by capping the mother centriole thereby preventing cilia formation (PubMed:17719545 PubMed:17681131, PubMed:23486064). Also involved in promoting ciliogenesis. May play a role in the assembly of the mother centriole subdistal appendages (SDA) thereby effecting the fusion of recycling endosomes to basal bodies during cilia formation. Required for correct spindle formation and has a role in regulating cytokinesis and genome stability via cooperation with CALM1 and CETN2 (PubMed:16760425). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9738
Species Human
Purification Method purified
Accession Number O43303
Common Name CP110
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 6330503K22Rik; AA415922; AI427129; AW557948; CCP110; centriolar coiled coil protein 110; centriolar coiled coil protein 110kDa; centriolar coiled-coil protein 110; centriolar coiled-coil protein 110kDa; centriolar coiled-coil protein of 110 kDa; Centriolin; centrosomal protein CP110; centrosomal protein of 110 kDa; CEP1; Cep110; CNTRL; CP110; FAN; Kiaa0419; RGD1310509
Gene Symbol CCP110
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.