Learn More
BIOHIPPO Human CCR3 Microsample
SDP
Catalog No. 5000314251
Encompass
Human CCR3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CCR3 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CCR3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CCR3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CCR3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14251 Supplier BIOHIPPO Supplier No. BHE15211854
Human CCR3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CCR3 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CCR3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CCR3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CCR3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color