Learn More
ACROBiosystems ACROBiosystems Human CCR5 Full Length Protein-VLP (HEK293)
Catalog No. 502106963
Encompass_Preferred
Recombinant Protein;100UG;Human CCR5 Full Length Protein-VLP (CC5-H52P3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Leu 352 (Accession # P51681-1).;This protein carries no "tag". The protein has a calculated MW of 40.5 kDa.
Catalog No. 50-210-6963 Supplier ACROBiosystems Supplier No. CC5H52P3100UG
Recombinant Protein;100UG;Human CCR5 Full Length Protein-VLP (CC5-H52P3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Leu 352 (Accession # P51681-1).;This protein carries no "tag". The protein has a calculated MW of 40.5 kDa.