Learn More
Sino Biological Recombinant Human CCR8 Protein (VLP, Flag & His-Strep Tag)
Catalog No. 502993419
Encompass_Preferred
A DNA sequence encoding the human CCR8 (P51685)(Met1-Ser318) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus. Within the expressed regions, there are several proprietary thermostabilizing mutations.This protein was prepared using our VLP platform. As an innovative technology platform, VLPs are generated through the self-assembly of viral proteins in HEK293 cells. This process efficiently integrates membrane proteins onto the VLP surface, preserving their native conformation and yielding soluble, high-concentration proteins that are ideally suited for immunization and antibody screening.
Catalog No. 50-299-3419 Supplier Sino Biological Supplier No. 11450H82H100
A DNA sequence encoding the human CCR8 (P51685)(Met1-Ser318) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus. Within the expressed regions, there are several proprietary thermostabilizing mutations.This protein was prepared using our VLP platform. As an innovative technology platform, VLPs are generated through the self-assembly of viral proteins in HEK293 cells. This process efficiently integrates membrane proteins onto the VLP surface, preserving their native conformation and yielding soluble, high-concentration proteins that are ideally suited for immunization and antibody screening.