Learn More
ACROBiosystems Human CCR8 2Vials
Catalog No. 502535264
Encompass_Preferred
This entry represents CC chemokine receptor 8 (CCR8) which it is expressed predominantly in lymphoid tissues and has also been found in glomerular podocytes and human umbilical vein endothelial cells (HUVECs). CCR8 is associated with Th2 lymphocytes which are critical for allergy and has a role in lymphocyte activation migration proliferation and differentiation and in allergic diseases. CCR8 binds to CCL1 (also known as I-309) and to CCL16 (also known as liver expressed chemokine). It also exhibits a high affinity for three chemokines of viral origin: vMIP-I vMIP-II and vMCC-I.
Catalog No. 50-253-5264 Supplier ACROBiosystems Supplier No. CHEKATP1402VIALS
This entry represents CC chemokine receptor 8 (CCR8) which it is expressed predominantly in lymphoid tissues and has also been found in glomerular podocytes and human umbilical vein endothelial cells (HUVECs). CCR8 is associated with Th2 lymphocytes which are critical for allergy and has a role in lymphocyte activation migration proliferation and differentiation and in allergic diseases. CCR8 binds to CCL1 (also known as I-309) and to CCL16 (also known as liver expressed chemokine). It also exhibits a high affinity for three chemokines of viral origin: vMIP-I vMIP-II and vMCC-I.