Learn More
Sino Biological Recombinant Human CCR9 Protein (VLP, Full-length, Flag & His-Strep Tag)
Catalog No. 502993506
Encompass_Preferred
A DNA sequence encoding the Human CCR9 (P51686) (Met1-Leu369) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus.
Catalog No. 50-299-3506 Supplier Sino Biological Supplier No. 30150H82H20
A DNA sequence encoding the Human CCR9 (P51686) (Met1-Leu369) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus.