Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human CCR9 Full-Length Protein (VLP), Thr 2-Leu 369, HEK293, 500 μg

Catalog No. NC3886316
Encompass_Preferred

Recombinant full-length human CCR9 is supplied as virus-like particles produced in HEK293 cells, preserving the native conformation of this seven-transmembrane GPCR for binding and functional assays.

  • Expressed in human HEK293 cells.
  • Covers amino acids Thr 2-Leu 369 (UniProt P51686-1).
  • Presented in virus-like particles to preserve native conformation.
  • Pack size: 500 μg (250 μg x 2).
  • Supplied in PBS and arginine, pH 7.4, with trehalose; 0.2 μm filtered.
  • Endotoxin < 1.0 EU/μg (LAL / rFC).
  • DLS mean peak radius 60-85 nm (>95% intensity).
  • QC-tested by ELISA: immobilized at 5 μg/mL binds anti-CCR9 antibody (linear 0.001-0.25 μg/mL).
  • Predicted N-terminus: Asp.

Catalog No. NC3886316 Supplier ACROBiosystems Supplier No. CC9H52P3500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human CCR9 is supplied as virus-like particles produced in HEK293 cells, preserving the native conformation of this seven-transmembrane GPCR for binding and functional assays.

  • Expressed in human HEK293 cells.
  • Covers amino acids Thr 2-Leu 369 (UniProt P51686-1).
  • Presented in virus-like particles to preserve native conformation.
  • Pack size: 500 μg (250 μg x 2).
  • Supplied in PBS and arginine, pH 7.4, with trehalose; 0.2 μm filtered.
  • Endotoxin < 1.0 EU/μg (LAL / rFC).
  • DLS mean peak radius 60-85 nm (>95% intensity).
  • QC-tested by ELISA: immobilized at 5 μg/mL binds anti-CCR9 antibody (linear 0.001-0.25 μg/mL).
  • Predicted N-terminus: Asp.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.