Learn More
TRIBIOSCIENCE INC Human CD30/TNFRSF18 ELISA, 1 x 96-well plate
SDP
Catalog No. NC3454586
Encompass_Preferred
Tribioscience’s Human CD30 Fast ELISA is designed to quantitatively detect human CD30 levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 7 to 2000 pg/mL.
Catalog No. NC3454586 Supplier TRIBIOSCIENCE INC Supplier No. TBS32109
Tribioscience’s Human CD30 Fast ELISA is designed to quantitatively detect human CD30 levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 7 to 2000 pg/mL.