Learn More
Biotang Inc Human CD112 / Nectin-2 / PVRL2 Protein (His Tag), 50ug, the extracellular domain (Met 1-Leu 360) of human CD112 (NP_002847.1) was expressed in human cells with a C-terminal polyhistidine tag.
Supplier:  Biotang Inc RPR9865
Encompass_Preferred
Human CD112 / Nectin-2 / PVRL2 Protein (His Tag), 50ug, the extracellular domain (Met 1-Leu 360) of human CD112 (NP_002847.1) was expressed in human cells with a C-terminal polyhistidine tag.
Catalog No. 50-751-5374