Learn More
Biotang Inc Human CD112 / Nectin-2 / PVRL2 Protein, 50ug, The mature form of recombinant human CD112 (NP_002847.1) extracellular domain (Met 1-Leu 360) with quinary-aa peptide (DDDDK) at the C-terminus was expressed and purified in human cells.
Supplier:  Biotang Inc RPR9866
Encompass_Preferred
Human CD112 / Nectin-2 / PVRL2 Protein, 50ug, The mature form of recombinant human CD112 (NP_002847.1) extracellular domain (Met 1-Leu 360) with quinary-aa peptide (DDDDK) at the C-terminus was expressed and purified in human cells.
Catalog No. 50-751-5373