Learn More
ACROBiosystems ACROBiosystems Human CD117 / c-kit Protein, His Tag
Catalog No. 502106571
Encompass_Preferred
Recombinant Protein;1MG;Human CD117, His Tag (CD7-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession # P10721-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.1 kDa. The protein migrates as 66-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD117, His Tag (Cat. No. CD7-H52H4) at 2 μg/mL (100 μL/well) can bind Biotinylated Human SCF, Avitag,His Tag (Cat. No. SCF-H82E1) with a linear range of 0.5-8 ng/mL (QC tested).
Catalog No. 50-210-6571 Supplier ACROBiosystems Supplier No. CD7H52H41MG
Recombinant Protein;1MG;Human CD117, His Tag (CD7-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession # P10721-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.1 kDa. The protein migrates as 66-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD117, His Tag (Cat. No. CD7-H52H4) at 2 μg/mL (100 μL/well) can bind Biotinylated Human SCF, Avitag,His Tag (Cat. No. SCF-H82E1) with a linear range of 0.5-8 ng/mL (QC tested).