Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems CD133 Protein, Full Length, VLP, 20 μg

Catalog No. 502331922
Encompass_Preferred

This product consists of full-length human CD133 (Prominin-1) protein presented as virus-like particles (VLPs). CD133 is a pentaspan membrane glycoprotein that serves as a marker for cancer stem cells in various malignancies, including glioblastoma, colorectal, and liver cancers. The protein is expressed in human 293 cells (HEK293) and incorporates transmembrane proteins into a lipid bilayer, preserving authentic conformation, post-translational modifications, and orientation in a detergent-free environment.

  • Greater than 95% purity as determined by SEC-HPLC
  • Contains amino acids Gly 20 to His 865
  • Endotoxin level is less than 1.0 EU per μg
  • Suitable for immunization, ELISA screening, and cell-based assays like FACS
  • Supplied as a 0.2 μm filtered solution in PBS, pH 7.4, with trehalose

Catalog No. 50-233-1922 Supplier ACROBiosystems Supplier No. CD3H52P320UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product consists of full-length human CD133 (Prominin-1) protein presented as virus-like particles (VLPs). CD133 is a pentaspan membrane glycoprotein that serves as a marker for cancer stem cells in various malignancies, including glioblastoma, colorectal, and liver cancers. The protein is expressed in human 293 cells (HEK293) and incorporates transmembrane proteins into a lipid bilayer, preserving authentic conformation, post-translational modifications, and orientation in a detergent-free environment.

  • Greater than 95% purity as determined by SEC-HPLC
  • Contains amino acids Gly 20 to His 865
  • Endotoxin level is less than 1.0 EU per μg
  • Suitable for immunization, ELISA screening, and cell-based assays like FACS
  • Supplied as a 0.2 μm filtered solution in PBS, pH 7.4, with trehalose

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.