Learn More
Sino Biological Human CD14 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 12078R00120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD14 (rh CD14; Catalog#10073-H08H; NP_000582.1; Met1-Met344).
Catalog No. 50-237-4263