Learn More
Sino Biological Recombinant Human PSGL-1/CD162 Protein (mFc Tag)
Supplier:  Sino Biological 10490H05H500
Encompass_Preferred
A DNA sequence encoding the human SELPLG (AAC50061.1) (Met1-Val295) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-222-3544