Learn More
Sino Biological Human CD146/MCAM Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786620
Encompass_Preferred
The human CD146 ELISA Pair Set is for the quantitative determination of human CD146.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD146(MCAM / MUC18) coated on a 96-well plate. Standards and samples are added tothe wells, and any CD146 present binds to the immobilized antibody. The wellsare washed and a horseradish peroxidase conjugated rabbit anti-CD146 polyclonalantibody is then added, producing an antibody-antigen-antibody "sandwich". Thewells are again washed and TMB substrate solution is loaded, which producescolor in proportion to the amount of CD146 present in the sample. To end theenzyme reaction, the stop solution is added and absorbances of the microwellare read at 450 nm.
Catalog No. 50-007-86620 Supplier Sino Biological Supplier No. SEK101155
The human CD146 ELISA Pair Set is for the quantitative determination of human CD146.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD146(MCAM / MUC18) coated on a 96-well plate. Standards and samples are added tothe wells, and any CD146 present binds to the immobilized antibody. The wellsare washed and a horseradish peroxidase conjugated rabbit anti-CD146 polyclonalantibody is then added, producing an antibody-antigen-antibody "sandwich". Thewells are again washed and TMB substrate solution is loaded, which producescolor in proportion to the amount of CD146 present in the sample. To end theenzyme reaction, the stop solution is added and absorbances of the microwellare read at 450 nm.