Learn More
Biotang Inc Human CD147 / EMMPRIN / Basigin Protein, 50ug, The mature form of human CD147 (NP_940991.1) extracellular domain (Met 1-His 205) with quinary-aa peptide (DDDDK) at the C-terminus was expressed and purified.
Supplier:  Biotang Inc RPR9844
Encompass_Preferred
Human CD147 / EMMPRIN / Basigin Protein, 50ug, The mature form of human CD147 (NP_940991.1) extracellular domain (Met 1-His 205) with quinary-aa peptide (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-751-5384