Learn More
Biotang Inc Human CD147 / EMMPRIN / Basigin Protein (His Tag), 50ug, the extracellular domain (Met 1-His 205) of human CD147 (NP_940991.1) precursor was expressed with the fused polyhistidine tag at the C-terminus.
Supplier:  Biotang Inc RPR9845
Encompass_Preferred
Human CD147 / EMMPRIN / Basigin Protein (His Tag), 50ug, the extracellular domain (Met 1-His 205) of human CD147 (NP_940991.1) precursor was expressed with the fused polyhistidine tag at the C-terminus.
Catalog No. 50-751-5383