Learn More
Invitrogen™ Human CD147/EMMPRIN ELISA Kit
Description
Human CD147/EMMPRIN quantitates human CD147/EMMPRIN in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CD147/EMMPRIN.
Specifications
Specifications
| Accession Number | 5A11; 5A11/Basigin; 5F7; AI115436; AI325119; Basic immunoglobulin superfamily; basigin; basigin (Ok blood group); Basigin (O x 47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); basignin; Basignin (O x 47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); Bsg; CD147; CE9; collagenase stimulatory factor; EMMPRIN; Extracellular matrix metalloproteinase inducer; Glycoprotein CE9; gp 42; HT7; HT-7; HT7 antigen; Leukocyte activation antigen M6; Membrane glycoprotein gp42; neurothelin; OK; OK blood group antigen; OX-47 antigen; O x 47 R; TCSF; Tumor cell-derived collagenase stimulatory factor; UNQ6505/PRO21383 |
| Assay Range | 12 to 2,500 pg/mL |
| Assay Sensitivity | 12 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 682 |
| Gene Symbol | BSG |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.