Learn More
Raybiotech Inc Human CD147/EMMPRIN ELISA Kit
Supplier:  Raybiotech Inc ELHCD1471
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 12 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human CD147.
Catalog No. 50-194-9071