Learn More
ANCELL CORPORATION Human CD152 CTLA-4 Fusion Protein, Immune Checkpoint Regulator, High-Affinity Binding, 25 µg
Supplier:  ANCELL CORPORATION 501820
Encompass
The ANCELL CORPORATION Human CD152 CTLA-4 (25 µg) is a potent negative regulator of T cell activation, designed as a soluble dimeric fusion protein. It exhibits high-affinity binding to CD80 and CD86, making it an essential tool for researchers studying immune checkpoint interactions. This product aids in investigating T cell signaling pathways and immune regulation, distinguishing it from other offerings with its optimized format for various in vitro assays.
Catalog No. NC0739531