Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CD155 (PVR) Fc Chimera Recombinant Protein

Catalog No. PIRP87969 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP87969 1 mg
PIRP87968 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP87969 Supplier Invitrogen™ Supplier No. RP87969
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (1205 μL) be added to the vial to prepare a stock solution of 0.83 mg/mL. Concentration is measured by UV.

The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15151
For Use With (Application) Binding Assay, Control, ELISA Standard
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 5817
Molecular Weight (g/mol) 61.8 kDa
Name Human CD155 (PVR) Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 1 mg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias 3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4
Common Name CD155
Gene Symbol PVR
Biological Activity 1. Measured by its binding ability in a functional ELISA. Immobilized human CD226 His at 2 μg/mL (100 μL/well) can bind human CD155 hFc, The EC50 of bind human CD155 hFc is 100-800 ng/mL 2. Immobilized TIGIT Protein, Human, Recombinant (ECD, His Tag) (at 2 μg/ml (100 μL/well) can bind CD155/PVR Protein, Human, Recombinant (ECD, hFc Tag), the EC50 of CD155/PVR Protein, Human, Recombinant (ECD, hFc Tag) is 25-140 ng/mL)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Human CD155, amino acids Met1-Asn343 (Accession # P15151-1) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity ELISA Activity: Immobilized Recombinant Human TIGIT / VSTM3 Protein (His Tag) (Cat: 10917-H08H3) at 2ug/mL (100uL/well) can bind Recombinant Human CD155/PVR/NECL5 Protein (Fc Tag) (Cat: 10109--H02H), the EC50 is 10-30ng/mL
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95 % as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.