Learn More
Abcam Human CD16 ELISA Kit- Intracellular, 1 x 96 Tests
Catalog No. 502711628
Encompass_Preferred
Human CD16 ELISA Kit- Intracellular is a single-wash 90-min Simplestep used to quantify Human CD16 with a sensitivity of 5.717 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-1628 Supplier Abcam Supplier No. AB3169101X96TESTS
Human CD16 ELISA Kit- Intracellular is a single-wash 90-min Simplestep used to quantify Human CD16 with a sensitivity of 5.717 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement