Learn More
Sino Biological Recombinant Anti-CD160 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12191R02320
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD160 / NK1 / BY55 (rh CD160 / NK1 / BY55; Catalog#12191-H08H; NP_008984.1; Met1-Leu158).
Catalog No. 50-255-4417