Learn More
Sino Biological Anti-CD163 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 310222T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CD163 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-9449