Learn More
Sino Biological Anti-CD164 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 12260MM05A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD164 (rh CD164; Catalog#12260-H08H; Q04900-1; Met1-Asp162) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-2328